Corning® Cell Culture Roller Bottles, Corning
Supplier: Corning
Certificates
About this item
Virgin PS, sterile.
- One piece seamless construction
- All bottles have printed lot numbers to aid product traceability
- Sterilized by gamma radiation
- Certified non-pyrogenic
- Ribbed design provides twice the surface area with the same exterior dimensions
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. All bottles are graduated, unless otherwise stated.
Note: Corning recommends 0.2 to 0.3 ml of medium per cm² of growth area, and setting roller rack speeds to provide 0.5 to 1.0 rpm.
Specifications
- Catalog No:
- 33500-694
- 89089-838
- 62404-370
- 33502-074
- 25382-462
- 46610-074
- 22250-132
- 62404-446
- 62404-444
- 62404-448
- 21008-662
- 21008-660
- 62404-442
- 62404-440
- 29442-914
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta