Synthware Vacuum/Gas Adapter, Ball Joint, Kemtech America
60000-170
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:Synthware Vacuum/Gas Adapter
- Environmentally Preferable:
- Cat. No.:60000-170
- Supplier no.:A293520
Vacuum/gas adapter with a ball joint.
- [ST] ball joint
- With hose connection
Featuring a [ST] 35/20 ball joint and 8mm hose connection.