To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:C/EBP epsilon
- Antigen Symbol:CEBPE
- Clonality:Monoclonal
- Clone:PCRP-CEBPE-1G12
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q15744
- Antigen Synonyms:CRP1|CCAAT/enhancer-binding protein epsilon|CCAAT/enhancer binding protein (C/EBP) epsilon|CEBPE|CEBPE_HUMAN|CCAAT/enhancer binding protein epsilon|CRP 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:32 kDa/30 kDa/27 kDa/14 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human CEBPE protein.
- Purification:Protein A/G chromatography
- Cat. No.:77179-244
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-CEBPE-1G12] antibody to CEBPE for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml
Type: Primary
Antigen: CEBPE
Clonality: Monoclonal
Clone: PCRP-CEBPE-1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human