About this item
Rabbit monoclonal [ARC5003-10] antibody to N6-Methyladenosine / m6A for ICC/IF, IP, RIP, Nucleotide Array and Dot Blot with samples derived from Species Independent.
- Validated Applications: ICC/IF, IP, RIP, Nucleotide Array, Dot Blot
- Recommended Dilutions: DB: 1:500 to 1:2,000, meRIP: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: N6-methyladenosine / m6A
Clonality: Monoclonal
Clone: ARC5003-10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Species Independent
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77215-098
- Antigen Symbol:
- N6-methyladenosine / m6A
- Antigen Name:
- N6-methyladenosine / m6A
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC5003-10
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Immunogen:
- Chemical compounds corresponding to N6-methyladenosine / m6A.
- Form:
- Liquid
- Purification:
- Protein A/G purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF, RIP, Dot Blot