To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:A32 antigen
- Antigen Symbol:CD146
- Clonality:Monoclonal
- Clone:MCAM/3179
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2c kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P43121
- Antigen Synonyms:Melanoma associated antigen A32|Cell surface glycoprotein P1H12|Mcam|Melanoma adhesion molecule|CD146 antigen|Cell surface glycoprotein MUC18|CD 146|Gicerin|CD146
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:130 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human MCAM protein
- Purification:Protein A/G chromatography
- Cat. No.:76963-876
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MCAM/3179] antibody to CD146 for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: MCAM/3179
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human