To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Arc 1
- Antigen Symbol:E Cadherin
- Clonality:Monoclonal
- Clone:67A4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P12830
- Antigen Synonyms:CD324|cadherin 1 type 1 E-cadherin|CD324 antigen|CAM 120/80|CD 324|Cadherin1|CADH1_HUMAN|Cadherin 1|CD 234
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:T-47D cells
- Tested Applications:IHC-Fr, ICC
- Purification:Protein A chromatography
- Cat. No.:76962-614
- Packaging:Plastic vial
Mouse monoclonal [67A4] antibody to E Cadherin for Flow Cytometry, IP, WB, IHC-Fr and ICC with samples derived from Human.
- Validated Applications: Flow Cytometry, IP, WB, IHC-Fr, ICC
- Recommended Dilutions: IHC-Fr: 4-8 µg/ml, Flow Cytometry: 5-10 µg/ml
Type: Primary
Antigen: E Cadherin
Clonality: Monoclonal
Clone: 67A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human