To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cambridge pathology 1 antigen
- Antigen Symbol:CD52
- Clonality:Monoclonal
- Clone:HI186
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 T
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P31358
- Antigen Synonyms:CD 52|CD52 antigen|CAMPATH 1|CD52_HUMAN|CAMPATH-1 antigen|CD52|CDw52|CD52 molecule|CAMPATH 1 antigen
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human tonsil
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76955-744
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [HI186] antibody to CD52 (FITC) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human