About this item
Mouse monoclonal (ABT-Calret 1) antibody to Calretinin for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: Calretinin
Clonality: Monoclonal
Clone: ABT-Calret 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77055-298
- 77055-300
- Antigen Symbol:
- Calretinin
- Calretinin
- Antigen Name:
- 29 kDa calbindin
- 29 kDa calbindin
- Antigen Synonyms:
- Calbindin 2 29kDa|CAL 2|CALB 2|CAB29|CALB2_HUMAN|CAB 29|Calbindin 2|CAL2|CALB2
- Calbindin 2 29kDa|CAL 2|CALB 2|CAB29|CALB2_HUMAN|CAB 29|Calbindin 2|CAL2|CALB2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-Calret 1
- ABT-Calret 1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P22676
- UniprotID# P22676
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Calretinin
- Synthetic peptide derived from human Calretinin
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage Buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC
- IHC