Anti-ALKBH7 Rabbit Polyclonal Antibody
Catalog # 76757-212
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:AlkB homolog 7
- Antigen Symbol:ALKBH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:84266
- Antigen Synonyms:alpha-ketoglutarate-dependent dioxygenase alkB homolog 7|spermatogenesis-associated protein 11|alkylated DNA repair protein alkB homolog 7|UNQ6002|alkB|mitochondrial|SPATA11|alkylation repair homolog 7|probable alpha-ketoglutarate-dependent dioxygenase ABH7|ABH7|spermatogenesis cell proliferation related protein|spermatogenesis associated 11
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human ALKBH7 (NP_115682.1).
- Purification:Affinity purification
- Cat. No.:76757-212
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ALKBH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat