Anti-ACSL5 Rabbit Polyclonal Antibody
50 µl, Vial
Catalog # 76762-948
Supplier: Prosci
Supplier #: 14-586
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: ACSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:50 µl
- Packaging:Vial
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification.
- Form:Liquid
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Isotype:IgG
- Host:Rabbit
- Concentration:Batch dependent
- Cat. No.:76762-948
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:20
- Antibody Type:Primary
- Antigen Symbol:ACSL5
- Gene ID:51703
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 500-739 of human ACSL5 (NP_057318.2).
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:ACS5|UNQ633/PRO1250|fatty-acid-Coenzyme A ligase|long-chain 5|long-chain acyl-CoA synthetase 5|long-chain fatty acid coenzyme A ligase 5|ACSL5|FACL5|ACS2|HGNC:16526|acyl-CoA synthetase long-chain family member 5
- Antigen Name:acyl-CoA synthetase long-chain family member 5
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



