Anti-KCNE1 Rabbit Polyclonal Antibody
Catalog # 76759-674
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:potassium voltage-gated channel subfamily E regulatory subunit 1
- Antigen Symbol:KCNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3753
- Antigen Synonyms:LQT2/5|LQT5|potassium voltage-gated channel|voltage gated potassiun channel accessory subunit|ISK|Isk-related family|MinK|voltage gated subfamily E regulatory beta subunit 1|JLNS2|Isk-related subfamily|potassium channel|minimal potassium channel|JLNS|delayed rectifier potassium channel subunit IsK|member 1|IKs producing slow voltage-gated potassium channel subunit beta Mink|potassium voltage-gated channel subfamily E member 1|cardiac delayed rectifier potassium channel protein
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KCNE1 (NP_000210.2).
- Purification:Affinity purification.
- Cat. No.:76759-674
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KCNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse