Anti-ALKBH5 Rabbit Polyclonal Antibody
76758-768
: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:alkB homolog 5, RNA demethylase
- Antigen Symbol:ALKBH5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:54890
- Antigen Synonyms:alpha-ketoglutarate-dependent dioxygenase alkB homolog 5|alkylation repair homolog 5|ABH5|OFOXD|RNA demethylase ALKBH5|alkB|probable alpha-ketoglutarate-dependent dioxygenase ABH5|AlkB family member 5|OFOXD1|RNA demethylase|oxoglutarate and iron-dependent oxygenase domain containing|alkylated DNA repair protein alkB homolog 5
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 245-394 of human ALKBH5 (NP_060228.3).
- Purification:Affinity purification.
- Cat. No.:76758-768
- Packaging:Vial
Type: Primary
Antigen: ALKBH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human