Anti-PACS2 Rabbit Polyclonal Antibody
Catalog # 77125-296
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphofurin acidic cluster sorting protein 2
- Antigen Symbol:PACS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:23241
- Antigen Synonyms:PACS1-like protein|PACS1L|Phosphofurin acidic cluster sorting protein 2|PACS-2
- Modification:None
- Storage Buffer:PACS2 antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:PACS2 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:PACS2 antibody was raised against a 17 amino acid peptide near the center of human PACS2.The immunogen is located within amino acids 330-380 of PACS2.
- Purification:PACS2 antibody is affinity chromatography purified via peptide column.
- Cat. No.:77125-296
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PACS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat