Anti-USP25 Rabbit Polyclonal Antibody
Catalog # 77126-512
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ubiquitin specific peptidase 25
- Antigen Symbol:USP25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:29761
- Antigen Synonyms:Deubiquitinating enzyme 25|USP25 Antibody: USP21|USP21|Ubiquitin carboxyl-terminal hydrolase 25
- Modification:None
- Storage Buffer:USP25 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:USP25 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:USP25 antibody was raised against a 17 amino acid peptide near the center of human USP25.The immunogen is located within amino acids 470-520 of USP25.
- Purification:USP25 antibody is affinity chromatography purified via peptide column.
- Cat. No.:77126-512
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: USP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse