Human Recombinant Vitronectin Protein (Animal Free) (from HEK293 cells)
77129-460
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Environmentally Preferable:
- Storage Conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological Activity:Recombinant human Vitronectin promotes attachment of hESC and iPSC in serum-free, feeder conditions at 5 µg/ml.
- Gene ID:7448
- Protein Synonyms:Serum-spreading factor|VTN|V75
- Protein/Peptide Name:VTN
- Purity:≥95% by SDS-PAGE gel and HPLC analyses
- Sequence:DQESCKGRCT EGFNVDKKCQ CDELCSYYQS CCTDYTAECK PQVTRGDVFT MPEDEYTVYD DGEEKNNATV HEQVGGPSLT SDLQAQSKGN PEQTPVLKPE EEAPAPEVGA SKPEGIDSRP ETLHPGRPQP PAEEELCSGK PFDAFTDLKN GSLFAFRGQY CYELDEKAVR PGYPKLIRDV WGIEGPIDAA FTRINCQGKT YLFKGSQYWR FEDGVLDPDY PRNISDGFDG IPDNVDAALA LPAHSYSGRE RVYFFKGKQY WEYQFQHQPS QEECEGSSLS AVFEHFAMMQ RDSWEDIFEL LFWGRTSAGT RQPQFISRDW HGVPGQVDAA MAGRIYISGM APRPSLAKKQ RFRHRNRKGY RSQRGHSRGR NQNSRRPSRA TWLSLFSSEE SNLGANNYDD YRMDWLVPAT CEPIQSVFFF SGDKYYRVNL RTRRVDTVDP PYPRSIAQYW LGCPAPGHL
- Formulation:Lyophilized
- Shipping Temperature:RT
- Cat. No.:77129-460