Mouse Recombinant R-Spondin-1 Protein (from CHO cells)
Catalog # 77126-462
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:5 µg
- Environmentally Preferable:
- Storage Conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological Activity:R-Spondin-1 enhances BMP-2-mediated differentiation of MC3T3-E1 cells.
- Gene ID:192199
- Protein Synonyms:Roof plate-specific Spondin-1|RSPO1
- Protein/Peptide Name:Rspo1
- Purity:≥95% by SDS-PAGE gel and HPLC analyses
- Sequence:SRGIKGKRQR RISAEGSQAC AKGCELCSEV NGCLKCSPKL FILLERNDIR QVGVCLPSCP PGYFDARNPD MNKCIKCKIE HCEACFSHNF CTKCQEGLYL HKGRCYPACP EGSTAANSTM ECGSPAQCEM SEWSPWGPCS KKRKLCGFRK GSEERTRRVL HAPGGDHTTC SDTKETRKCT VRRTPCPEGQ KRRKGGQGRR ENANRHPARK NSKEPGSNSR RHKGQQQPQP GTTGPLTSVG PTWAQ
- Formulation:Lyophilized
- Shipping Temperature:RT
- Cat. No.:77126-462