Anti-ENPP2/AUTOTAXIN Goat Polyclonal Antibody
Catalog # 77121-790
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ectonucleotide pyrophosphatase/phosphodiesterase 2
- Antigen Symbol:ENPP2/AUTOTAXIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5168
- Antigen Synonyms:extracellular lysophospholipase D|ATX|AUTOTAXIN|autotaxin-t|ENPP2|ectonucleotide pyrophosphatase/phosphodiesterase 2|phosphodiesterase|LysoPLD|E-NPP 2|ATX-X|ectonucleotide pyrophosphatase/phosphodiesterase family member 2|NPP2|PD-IALPHA|PDNP2
- Modification:None
- Storage Buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage Temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping Temperature:20
- Immunogen:The immunogen for this antibody is: PDHLTSCVRPDVRVS.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77121-790
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ENPP2/AUTOTAXIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human