SARS-CoV-2 (COVID-19) Recombinant S RBD Protein (from Mammalian Cells)
77133-884
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Size:0.1 mg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week. Store at −20...−80 °C for twelve months from the date of receipt.
- Biological Activity:Recombinant SARS-CoV-2 (COVID-19) S Protein RBD (77133-884) can bind Anti-SARS-CoV-2 (S1)-1 antibody (76757-076) measured by a functional ELISA assay.The EC50 of Recombinant SARS-CoV-2 (COVID-19) S Protein RBD is 15 ng/ml.
- Protein Synonyms:2019-nCov RBD Protein|2019-nCoV Spike RBD Protein
- Protein/Peptide Name:S RBD
- Purity:>90% as determined by SDS-PAGE quantitative densitometry by Coomassie Blue Staining
- Molecular Weight:28.03 kDa
- Sequence:Arg319-Phe541
- Formulation:Lyophilized. Supplied as Lyophilized from PBS, pH 7.5. Please reconstitute the protein with ddH₂O to the concentration with 1 mg/ml.
- Shipping Temperature:RT
- Tested Applications:ELISA
- Cat. No.:77133-884