Order Entry
United States
ContactUsLinkComponent
Anti-ADRB2 Recombinant Antibody [Clone: R11/E1]
Anti-ADRB2 Recombinant Antibody [Clone: R11/E1]
Catalog # 76629-488
Supplier:  Prosci
Anti-ADRB2 Recombinant Antibody [Clone: R11/E1]
Catalog # 76629-488
Supplier:  Prosci
Supplier Number:  24-796
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    adrenoceptor beta 2
  • Antigen Symbol:
    ADRB2
  • Clonality:
    Recombinant
  • Clone:
    R11/E1
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.2 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    154
  • Antigen Synonyms:
    β-2 adrenoceptor|β2AR|R11E1|Beta-2 adrenergic receptor|Beta-2 adrenoreceptor|Beta-2 adrenoceptor|β-2 adrenergic receptor|β2-AR|β-2 adrenoreceptor
  • Modification:
    Unmodified
  • Storage Buffer:
    PBS with 0.02% Proclin 300
  • Storage Temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizing mice with beta-2 adrenergic receptor.
  • Cat. No.:
    76629-488
  • Packaging:
    Vial

Specifications

About this item

Type: Primary
Antigen: adrenoceptor beta 2
Clonality: Recombinant
Clone: R11/E1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human