Anti-CSF2RB Recombinant Antibody [Clone: BION-1]
76629-434
: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:colony stimulating factor 2 receptor subunit beta
- Antigen Symbol:CSF2RB
- Clonality:Recombinant
- Clone:BION-1
- Conjugation:Unconjugated
- Host:
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.2 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:1439
- Antigen Synonyms:IL3RB|IL-3R common beta subunit|SMDP5|IL5RB|Beta Common Chain|Common Beta-Chain|cytokine receptor common subunit beta|low-affinity (granulocyte-macrophage)|colony stimulating factor 2 receptor|beta|beta-c|CDw131|CD131|CSF2RB
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:BALB/c mice were immunized intraperitonally with COS cells transfected DQP (Domain 4 of the common beta-chain) expression constructs. The immunizations were repeated 4 times at 2-week intervals. Four weeks after the final immunization, a mouse was boosted with COS transfectants intravenously. Three days later, splenocytes were harvested and fused with NS-1 myeloma cells. Hybridoma supernatants were screened on CHO DQP cells by flow cytometry, with untransfected CHO cells as a control. All antibodies were from single hybridoma clones as selected by a limiting dilution method. MoAbs were purified from ascites fluid or hybridoma supernatant by a protein A sepharose column.
- Tested Applications:Crystallography,Blocking,Functional Assays
- Cat. No.:76629-434
- Packaging:Vial
Type: Primary
Antigen: colony stimulating factor 2 receptor subunit beta
Clonality: Recombinant
Clone: BION-1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human