ContactUsLinkComponent
Hydro AccuMax Dilution System, 1-Button Dilution, High Flow
Hydro AccuMax Dilution System, 1-Button Dilution, High Flow
Catalog # 77007-812
Supplier:  Unimed-Midwest
undefined
Hydro AccuMax Dilution System, 1-Button Dilution, High Flow
Catalog # 77007-812
Supplier:  Unimed-Midwest
Supplier Number:  HDSC412331
Specifications
  • Description:
    Hydro AccuMax dilution system, 1-button dilution, high flow carboy fill (3.5 gpm)
  • Environmentally Preferable:
  • Cat. No.:
    77007-812
Specifications
About this item
Systems using AccuPro pressure regulation technology eliminate dilution variance by controlling the water pressure into the unit, never allowing the dilution to 'lean out' the desired dilution target.
  • 1-button dilution: high flow carboy fill (3.5 gpm)
  • New, expansive workspace to easily route tubes
  • Easy, tool-free maintenance
  • E-gap and AirGap options to satisfy regulatory standards
  • Same mounting brackets as 'Hydro Systems' AccuPro and AccuDose
High water pressure results in improperly diluted chemicals, poor cleaning performance, and lost chemical sales. AccuPro technology delivers accurate dilution regardless of fluctuations in water pressure.