Specifications
- Description:PTS Collect™ capillary tubes, 30 μl
- Environmentally Preferable:
- Cat. No.:77020-358
Specifications
About this item
PTS Collect™ capillary tubes provide speed, efficiency and savings — especially for applications where exact sample volumes are required.
- Ease-of-use with no assembly
- One-hand blood collection and dosing
- Glass tube for easy blood collection
- Bulb-squeeze dispensing technology that ensures consistent performance
- Only one item to inventory (no plunger)
PTS Collect capillary tubes provide better efficiency and are designed to be easier to use than traditional blood transfer tubes. Using a single glass tube with plastic casing, PTS Collect capillary tubes provide consistent self-wicking technology to preset volumes. This improved design is perfect for blood collection needs covering a wide range of applications across many different medical devices.