ContactUsLinkComponent
BD MAX™ PCR Benchtop System
BD MAX™ PCR Benchtop System
  76808-680
 :  BD
undefined
BD MAX™ PCR Benchtop System
  76808-680
 :  BD
 :  441916
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    BD MAX™ PCR benchtop system
  • Environmentally Preferable:
  • Cat. No.:
    76808-680
The BD MAX™ System is a fully-integrated, automated platform that performs nucleic acid extraction and real-time PCR providing results for up to 24 samples across multiple syndromes in less than three hours.
  • Raise staff productivity through automation
  • Leverage a versatile menu and streamlined workflow to reduce total cost
  • Improve turnaround time for fast, appropriate treatment decisions
  • Address complex patient needs through unique assay design
  • Tailor testing to manage variable workloads
  • Provide practitioners and patients accurate, timely results
  • Guide patient therapy, management, and overall experience
  • Improve institutional, operational, and financial outcomes
With the BD MAX™ System, laboratories can leverage a broad, expanding suite of molecular solutions.