Storage Microplates, 96 Square Well, Deep Well, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
These storage plates are designed for high density sample collection and storage for a wide array of applications within cell biology, molecular biology and drug discovery. Clear plate, perfect for storage applications.
- Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
- Autoclavable and stackable
- Free from RNase, DNase, human genomic DNA and endotoxins
- Alphanumeric grid reference to aid well and sample identification
- SBS footprint
- Square wells, to make the best use of space, and to improve mixing
- Temperature range for use: –80 to 120 °C
- Suitable for adhesive and heat sealing, and as a collection plate from filter systems
V-Shaped bottom is best for precipitation, centrifugation and small volume recovery due to the V-shaped bottom, as the liquid gathers at the lowest point of the V for easy pipetting.
U-shaped bottom plates are best for washing, mixing and pelleting and give high surface area.
Accessories information: 96 square well sealing cap mat, clear silicone, for use with square 96 well microplates and deep well storage microplates.