To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Podoplanin
- Clonality:Monoclonal
- Clone:D2-40
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:6 ml
- Environmentally Preferable:
- Cat. No.:76634-530
Podoplanin is a 38 kDa membrane mucoprotein originally detected on the surface of rat podocytes .The antibody against podoplanin (D2-40) reacts to a 40 kDA sialogylcoprotein found on lymphatic endothelium, fetal testis, and on the surface of testicular germ cell tumors. Podoplanin can be used to assist pathologists in identifying lymphatic endothelium to determine lymphatic invasion and also can be used to help differentiate mesothelioma from adenocarcinoma.