ContactUsLinkComponent
Klercide™ Cleanroom Quat Unit Dose Concentrate
  76554-242
 :  Avantor
undefined
Klercide™ Cleanroom Quat Unit Dose Concentrate
  76554-242
 :  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    50x2.6oz
  • Environmentally Preferable:
  • Cat. No.:
    76554-242
  • Packaging:
    Tub, double bagged
  • Sterility:
    Sterile
  • Form:
    Unit dose concentrate
Klercide™ Cleanroom Quat Unit Dose Concentrate is a sterile blend of quaternary ammonium compound and Water for Injection (WFI).
  • Broad Spectrum Efficacy - Unit dose concentrate can be effectively used as a sanitizer, disinfectant and fungicide (depending on dilution) on hard, non-porous surfaces – formulated for use in cleanrooms, critical manufacturing environments and other controlled areas.
  • Minimal-Residue Quat Formulation when Diluted as Sanitizer - Active quaternary ammonium molecule is efficacious and has non-corrosive material compatibility when diluted as a sanitizer. It accumulates significantly less residue than other broad spectrum cleanroom biocides. This product is a non-food contact sanitizer for use on hard, non-porous surfaces, making it ideal for daily use in cleanrooms.
  • Minimize Residue Associated Risk - Non-phenolic formulation. Reduced residue mitigates slip-and-fall risks associated with sticky residue buildup.
  • Cleanroom-Grade Production - Solution is 0.2 micron-filtered and aseptically-filled under unidirectional Grade A / ISO Class 5 air-flow in a Grade B / ISO Class 5 cleanroom — and double-bagged to simplify movement into high-grade areas. Supported with Certificates of Analysis and Sterility with every batch.
For use in cleanrooms and controlled areas, Klercide™ Cleanroom Quat Unit Dose Concentrate is a sanitizer, disinfectant, and fungicide for use on hard, non-porous surfaces. The product is effective at low concentrations, resulting in minimal residues and residue accumulation. Depending on dilution, product can be used as either a sanitizer, disinfectant or fungicidal.
The product is 0.2 micron filtered and sterile filled under ISO Class 5 laminar air follow in an ISO Class 5 cleanroom, and double bagged in packs of 5.
EPA Registered Sanitizer, Disinfectant, and Fungicide.
EPA Reg. # 1677-264