Inert PFA Spray Chambers for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
PFA Scott-type spray chamber and end-cap for the inert PFA sample introduction kit for Agilent ICP-MS.
- PFA Scott-type spray chamber provides excellent sensitivity, stability and washout
- PFA end cap is O-ring free and features a 6 mm nebulizer port and a make up gas port
- Manufactured from the highest purity grade PFA resin for the lowest blank contribution, ensuring it is suitable for semiconductor applications
The inert PFA sample introduction kit for Agilent ICP-MS instruments features a Scott type spray chamber offering excellent sensitivity, stability and washout. The end cap is O-ring free and features a 6 mm nebulizer port and a make up gas port.;Manufactured from the highest purity grade PFA resin, the inert components maintain the lowest blank contribution from the sample introduction system, ensuring it is suitable for semiconductor applications.