Parker Hannifin Flare Reducing Union Fittings
: Avantor
Leak-tight and high-pressure connection for corrosive chemical applications.
- Constructed of durable PFA and PVDF
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
- Environmentally Preferable: