Human Recombinant respiratory syncytial virus (RSV) (A2) Fusion glycoprotein/RSV-F Protein (ECD, His Tag) (from Baculovirus Insect cells)
Catalog # 103621-774
Supplier: Sino Biological
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:RSV
- Size:100 µg
- Environmentally Preferable:
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P03420.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HRSVgp08| fusion protein|F
- Protein/Peptide Name:RSV-F
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant human RSV-F comprises 514 amino acids with a predicted molecular mass of 57.4 kDa. The RSV F0 precursor protein is cleaved into the disulfide-linked F1 and F2 subunits. As a result of glycosylation, the apparent molecular mass of the protein is approximately 45-55 kDa and 18 kDa in SDS-PAGE under reducing conditions, corresponding to the two subunits respectively.
- Sequence:Met1-Thr529
- Shipping Temperature:Ambient
- Cat. No.:103621-774
Specifications
About this item
A DNA sequence encoding the extracellular domain of human RSV Fusion glycoprotein (P03420.1) (Met1-Thr529) was expressed fused with a polyhistidine tag at the C-terminus.