Human Recombinant NGF/NGFB Protein (from E. coli)
Catalog # 103886-342
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Environmentally Preferable:
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:NP_002497.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HSAN5|Beta-NGF|NGFB
- Protein/Peptide Name:NGF
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:The recombinant human NGF consists of 224 amino acids and predicts a molecular mass of 25.1 kDa.
- Sequence:Glu19-Ala241
- Shipping Temperature:Ambient
- Cat. No.:103886-342
Specifications
About this item
A DNA sequence encoding the human NGF (NP_002497.2) (Glu19-Ala241) was expressed with an initial Met.