Order Entry
United States
ContactUsLinkComponent
 
Sus scrofa (Pig) Recombinant Transferrin/TF Protein (His Tag) (from HEK293 Cells)
Catalog # 103623-462
Supplier:  Sino Biological
CAS Number:  
undefined
Sus scrofa (Pig) Recombinant Transferrin/TF Protein (His Tag) (from HEK293 Cells)
Catalog # 103623-462
Supplier:  Sino Biological
Supplier Number:  13656-W08H-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Pig
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to bind human His-TFRC (cat: 11020-H07H) functional ELISA.
    2. Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is typically 0.01-0.04 μg/ml.
  • Gene ID:
    NP_001231582.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    PRO2086|HEL-S-71p|PRO1557|TFQTL1|Transferrin
  • Protein/Peptide Name:
    TF
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The secreted recombinant Sus scrofa (Pig) transferrin comprises 705 amino acids and has a predicted molecular mass of 78.3 kDa. It migrates as an approximately 72 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Thr715
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103623-462

Specifications

About this item

A DNA sequence encoding the pig (Sus scrofa) transferrin (NP_001231582.1) (Met 1-Thr 715) was fused with the a polyhistidine tag at the C-terminus.