Automated cell lysis through bead beating.
- High-throughput tissue processing
- Efficient vertical motion
- Programmable touch screen with pre-programmed protocols
- Compatible with Spex DNAmax nucleic acid extraction kits
The Spex® SamplePrep Genomax® is an automated high-throughput plant and animal tissue homogenizer and cell lyser designed for rapid cell disruption, cell lysis and tissue homogenization through bead beating. This enables fast and efficient extraction of nucleic acids, proteins and other molecules.
The Genomax® is programmed via touch screen interface allowing users to set and store run time, operating rate, pause time and cycles. Protocols can be passcode protected. The Genomax comes pre-loaded with one-touch protocols for plant and animal tissue homogenization.
DNA isolation kits are available for extraction and purification of DNA from animal and plant tissues. In addition, a full range of Kryo-Tech® accessories are available to preserve temperature sensitive samples.
Typical applications include molecular biology, pharmaceutical, research, and extraction.
Typical samples include plant tissue, animal tissue, cell cultures, and microorganisms.