Order Entry
United States
ContactUsLinkComponent
VWR® USB Digital Data Logger (DDL) (Single Probe)
VWR® USB Digital Data Logger (DDL) (Single Probe)
Catalog # 76520-152
Supplier:  VWR International
undefined
VWR® USB Digital Data Logger (DDL) (Single Probe)
Catalog # 76520-152
Supplier:  VWR International

Specifications

  • Interface:
  • Description:
    USB digital data logger (DDL) (single probe)
  • Environmentally Preferable:
  • Weight:
    1.1 kg (2.5 lbs.)
  • Exterior W×D×H:
    203×127×76 mm (8×5×3")
  • Cat. No.:
    76520-152

Specifications

About this item

This USB data logger is compliant with current CDC vaccine storage monitor guidelines. It has a buffered probe in product simulated solution, °F/°C switchable readout, a detachable single probe, audible and visual alarms, and temperature data downloadable via USB drive (not included). Device stores up to 3 years of data.

  • Material: Assorted materials