Borosil® Liebig Condensers with Drip Tip and Interchangeable Inner/Outer Joint and Jacket, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
Made of Borosilicate glass 3.3 to withstand heat and temperature changes.
- Clear color
- Class/quality grade Type I, Class A
Simple design with a low cooling capacity. The light wall of the inner tube facilitates efficient heat transfer. Specifically used for distillate separation.
Certifications: Comply with IS 2626.