A high performance garment that protects against airborne contamination and static charges.
- Engineered for use in Class 100 cleanrooms
- Knee-length
- Military collar
- ESD knit cuffs
- YKK zipper closure
Thes ESD Cleanroom Frock is engineered to meet up to Class 100 cleanroom demands in microelectronic, semi-conductor, disk drive, laser, and similar industries. These frocks are made with static dissipative fabric to eliminate static on the wearer in cleanroom environments.
All garment seams are designed to maintain electrical continuity from panel-to-panel and from sleeve-to-sleeve in accordance with the ESD Association Garment Standard, ESD-STM2.1. These knee-length frocks protect the environment from the number one source of contamination found in cleanrooms - people. The frock features a military collar, ESD knit cuffs, and YKK zipper closure with adjustment snaps at the neck. Meets or exceeds requirements of ANSI ESD-S20.20 and the recommendations of ESD 4.1.
- Catalog No:
- 76489-946
- 76489-944
- 76489-954
- 76489-952
- 76489-950
- 76489-948
- 76489-960
- 76489-958
- 76489-956
- Garment application:
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Industrial, Electronics/semi-conductor, Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure