Differential Scanning Calorimetry, DSC 4000, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
The DSC 4000 provides dependable performance and quality results. This single-furnace, heat flux DSC is designed to run all your routine applications and includes an easy-to-load vertical furnace that is resistant to oxygen and chemical corrosion.
- Integrated mass flow controller for convenient gas control and switching
- Fast 45-position autosampler option for reliable unattended operation
- Portable cooling device (PCD) for cost-effective operation at low temperature
- Efficient, easy-to-clean nickel chromium sample plate
- Temperature range: −100 to +450 °C
- Thermal analysis analytical technique
- Pyris Software and Pyris Player operating system
- Not portable
Typical applications for the DSC 4000 are QA/QC testing, undergraduate teaching practicals, evaluating additives for plastic lifetime optimization, ideal for oxidation induction test (OIT).