Melting Point Apparatus, METTLER TOLEDO
Supplier: Mettler Toledo
Certificates
About this item
Melting point determination has never been so easy - with simple operating routines and an intuitive color touch screen, measurements can be performed quickly.
- Everything needed for the simple, automatic determination of melting point
- One Click™ melting point determination
- Widescreen display with color touch screen
- Color video recording
- Intensity curves and results can be viewed directly on the color touch screen
- Capability to export data as PDF
This melting point systems offer simultaneous determination of samples, statistical analysis, and printable results. The outstanding temperature-accuracy specifications and the compliance with current standards such as Ph. Eur., USP and others ensure accurate results for melting point and melting range determination up to 300 °C.