About this item
Cyto-Chex® BCT is a direct draw blood collection tube that maintains cellular morphology and surface antigen expression, including cluster of differentiation (CD) markers prior to analysis by flow cytometry.
- Up to 14-day sample stability when analyzed by flow cytometry
- FDA 510(k) cleared for consistent recovery of HIV-associated lymphocyte subsets for 14 days
- Available in 2 and 5 ml vials
Whole blood samples collected in Cyto-Chex® BCT can be stored at room temperature for up to 14 days. Cyto-Chex® BCT has been FDA 510(k) cleared for consistent recovery of HIV-associated lymphocyte subsets (CD3, CD4, CD8, CD19, CD16/CD56 and CD45) for up to 14 days.