Order Entry
United States
ContactUsLinkComponent
Clorox® Clean-Up Cleaner with Bleach
Clorox® Clean-Up Cleaner with Bleach
Catalog # 76452-004
undefined
Clorox® Clean-Up Cleaner with Bleach
Catalog # 76452-004
Supplier Number:  CLO31221

Specifications

  • Description:
    Clorox® Clean-Up Cleaner with Bleach
  • Size:
    32 oz
  • Environmentally Preferable:
  • Cat. No.:
    76452-004
  • Packaging:
    Trigger spray bottle
  • Scent:
    Original

Specifications

About this item

Powerful and refreshing Clorox® Clean Up® Cleaner with Bleach is specially designed to quickly and effectively clean, disinfect and deodorize a variety of surfaces. Removes dirt, grease, grime, stains and viruses.

  • Effective formula that removes tough stains, dirt, grease and grime with a fresh fragrance
  • Disinfects and kills 99.9% of common household bacteria in only 30 seconds
  • Kills viruses that cause colds and flu
  • Eliminates odors caused by bacteria, mold and mildew
  • Features Smart Tube® Technology so you'll spray every drop
  • Great for hard, non-porous surfaces, including kitchen sinks, appliances, floors, showers and more
  • Ideal for use in restaurants, cafeterias, hotels and other commercial facilities

It removes tough stains, grease, dirt and kills 99.9% of germs while eliminating odors caused by bacteria, mold and mildew. It's great on hard, nonporous surfaces, including kitchen and bathroom sinks, counters, refrigerators, appliances, bathtubs, toilets, fiberglass, floors, showers and tiles. Smart Tube® Technology spray bottle lets you spray every last drop. Clorox® Clean-Up® Cleaner + Bleach leaves your house smelling fresh and sparkling clean.

Chemical compound: Sodium Hypochlorite