To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml (2000 ppm)
- Matrix:Methanol (P&T)
- Pack type:Ampoule amber
- Description:EDB/DBCP Analytes Mix (High Level) for US EPA Methods 504 and 8011
- Size:1 ml
- Environmentally Preferable:
- Cat. No.:BDH0723-1ML
Specifications
About this item
Volatiles standard.
US EPA Method 504 is an analytical method for the monitoring of 1,2-Dibromomethane (EDB), 1,2-Dibromo-3-Chloropropane (DBCP), and 1,2,3-Trichloropropane (123TCP) in water by micro-extraction and either GC/ECD or GC/MS techniques.
US EPA Method 8011 is an analytical method for the monitoring of EDB and DBCP in drinking water and ground water by GC/MS.