ContactUsLinkComponent
Lower Reactor Support for CHEMRXNHUB™ Triple Support Stands, Benchtop Reactors, Chemglass
Lower Reactor Support for CHEMRXNHUB™ Triple Support Stands, Benchtop Reactors, Chemglass
Catalog # 76333-370
Supplier:  Chemglass
undefined
Lower Reactor Support for CHEMRXNHUB™ Triple Support Stands, Benchtop Reactors, Chemglass
Catalog # 76333-370
Supplier:  Chemglass
Supplier Number:  CG-1949-X-33
Specifications
  • Description:
    Lower reactor support for triple stand, 2 and 5l vessel capacity, ptfe coated
  • Environmentally Preferable:
  • Cat. No.:
    76333-370