Order Entry
United States
ContactUsLinkComponent
Brady® COVID-19 Vinyl Sign Kit; Social Distancing Seat Check, Brady Worldwide
Brady® COVID-19 Vinyl Sign Kit; Social Distancing Seat Check, Brady Worldwide
Catalog # 76420-562
Supplier:  Avantor
undefined
Brady® COVID-19 Vinyl Sign Kit; Social Distancing Seat Check, Brady Worldwide
Catalog # 76420-562
Supplier:  Avantor

Specifications

  • Color:
    Green/Red
  • Description:
    B-581 Sign, Social Distancing Seat Markers, Covid-19
  • Height:
    6"
  • Material:
    Vinyl
  • Environmentally Preferable:
  • Cat. No.:
    76420-562
  • Depth:
    0.035"
  • Width:
    6"

Specifications

About this item

Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue.

  • Kit contains 15 "X" markers and 5 “check” markers
  • Equipped with corner holes for placement and mounting
  • Keep customers and employees safe with this product

Mark seats and tables with this repositionable marker kit. It provides a convenient, professional-looking solution for conveying appropriate social distancing.