The simplicity and convenience of Mix-n-Stain™ labeling, for conjugation of 1 mg IgG with fluorescent CF® dyes or other dyes.
- Rapid, simple, covalent antibody conjugation
- No purification required after labeling
- Reaction takes only 30 minutes, with minimal hands-on time
- Wide selection of next-generation CF® dyes with superior brightness and photostability
- One labeling reaction of 1 mg IgG per kit
Mix-n-Stain™ Maxi Antibody Labeling Kits allow you to rapidly label 1 mg of IgG with one of Biotium's next-generation CF® dyes or other fluorescent dyes. Simply mix your antibody with the reaction buffer and optimally formulated dye provided, followed by a 30 minutes incubation and addition of the provided stop buffer. Any remaining free dye or other label is no longer reactive at the end of the labeling, so the conjugate is ready for staining without further purification. After the reaction, the antibody will be labeled with an average of 4 to 6 dye molecules per antibody molecule. Mix-n-Stain™ labeling is covalent, so labeled antibodies can be used for multiplex staining without transfer of dyes/labels between antibodies.
These kits feature a choice of Biotium’s next-generation CF® dyes, which were designed for advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes. Learn more about CF® Dyes. We also offer kits for labeling with fluorescein (FITC), Cyanine 555 (structurally identical to Cy®3) or Cyanine 647 (structurally identical to Cy®5).
Alexa Fluor and DyLight are registered trademarks of Thermo Fisher Scientific. Cy Dye is a registered trademark of GE Healthcare.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Environmentally Preferable: