PolyPro-ST Nebulizer for PerkinElmer ICP-MS, PerkinElmer
Catalog # 10138-240
Supplier: PerkinElmer
Specifications
- Description:PolyPro-ST nebulizer for PerkinElmer ICP-MS
- Environmentally Preferable:
- Cat. No.:10138-240
- Supplier no.:N0777220
Specifications
About this item
This PolyPro-ST nebulizer with detachable capillary is used with ELAN® and NexION® ICP-MS series instruments.
- Polypropylene construction
- Hydrofluoric acid resistant
- 0 to 3 ml/min. sample flow rate with peristaltic pumping
- Resistant to clogging and accidental breakage
The PolyPro-ST Nebulizer is an HF resistant alternative to the PFA nebulizer, but has the same high efficiency aerosol generation. It is a robust nebulizer that can self-aspirate with an exchangeable external sample uptake capillary, or it can be used with a peristaltic pump.