Adjustable Torch Mount Assembly for ELAN® 9000/DRC-e, PerkinElmer
Adjustable torch mount assembly for ELAN® 9000/DRC-e
Catalog # 89260-246
Supplier: PerkinElmer
Supplier #: W1020668
Adjustable Torch Mount Assembly for ELAN® 9000/DRC-e, PerkinElmer
Catalog # 89260-246
Supplier: PerkinElmer
Jump to:
Product Details & Documents
This torch mount assembly is used with ELAN® 9000 and DRC-e ICP-MS series instruments.
The cassette torch mount is an innovative torch design that is fully modular, demountable, rugged, and easy to maintain. It does not include the demountable quartz torch or injector.
Specifications
- Description:Adjustable torch mount assembly for ELAN® 9000/DRC-e