About this item
Quickly deliver collaborative and productive comfort to your workday by seamlessly alternating between sitting and standing with the Luxor pneumatic adjustable height standing desk.
- High speed crank allows for rapid adjustment from sitting to standing
- Crank handle quickly raises the desk height between 32.6" and 48.5" (with casters)
- Crank handle quickly raises the desk height between 29.6" and 45.5" (with glides)
- Spacious 59×29.5" desktop provides plenty of room for multiple monitor setups
- Includes choice of leveling glides and 3" casters, two with locking brake
- Max weight capacity: 154 lbs.
Now available with a high-speed crank operated lift mechanism, the turn-handle crank quickly moves the desk from the lowest to highest positions within seconds without requiring electric power.
The generous 59×29.5" work surface easily fits triple monitors plus a full-sized keyboard and mouse while the height adjustment range accommodates taller and shorter users. Each desk includes a choice of leveling glides as well as 3" casters, two with locking brakes, allowing for easy maneuverability to suit ever-changing workspace needs.
Enjoy the health benefits of standing while increasing focus and productivity with a high-speed crank adjustable desk from Luxor.
Overall with casters: W59×D29.5×H32.6 - 48.5";
Overall with glides: W59×D29.5×H29.6 - 45.5";
Desktop: W59×D29.5"