Order Entry
United States
ContactUsLinkComponent
Bilingual No Top Freight Labels, Labelmaster
Bilingual No Top Freight Labels, Labelmaster
Catalog # 101377-054
Supplier:  Label Master
Bilingual No Top Freight Labels, Labelmaster
Catalog # 101377-054
Supplier:  Label Master
Supplier Number:  BLT88

Specifications

  • Description:
    Shipping labels
  • Dimensions:
    50.8×101.6 mm (2×4")
  • Material:
    Fluorescent orange paper
  • Thickness:
    7.1 mil.
  • Environmentally Preferable:
  • Cat. No.:
    101377-054
  • Legend:
    No Top Freight
  • Supplier no.:
    BLT88

Specifications

About this item

Ensure the safety of shipping pallets with Bilingual No Top Freight labels.

  • Bold lettering
  • Fluorescent orange ink
  • Printed in English and Spanish
  • Made of paper
  • Features permanent adhesive for excellent adhesion to cardboard boxes

These high-visibility ColorBrite™ labels command attention and help increase handler safety. They are ideal for alerting shipping and receiving personnel of special handling instructions.