Order Entry
United States
ContactUsLinkComponent
Premier® EHEC Kit, Meridian Bioscience
Premier® EHEC Kit, Meridian Bioscience
Catalog # 76317-618
Supplier:  Meridian
CAS Number:  
undefined
Premier® EHEC Kit, Meridian Bioscience
Catalog # 76317-618
Supplier:  Meridian
Supplier Number:  608096
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    Premier® EHEC kit
  • Environmentally Preferable:
  • Cat. No.:
    76317-618
  • No. of tests:
    96 wells

Specifications

About this item

The Premier® EHEC test is a rapid in vitro microwell EIA for the detection of Shiga toxins I and II (Verotoxins) in stool specimens, broth cultures, from individual colonies or colony sweeps of agar plates. This test is intended for use as an aid in the diagnosis of enterohemorrhagic E. coli (EHEC) infections.

This test utilizes monoclonal anti-Shiga toxin capture antibodies absorbed to microwells. Four diluted samples are added to the wells and incubated at room temperature. A wash is performed to remove unbound material. Polyclonal anti-Shiga-like toxin antibodies are added for detection and incubated at room temperature. Another wash is used to remove unbound antibody. Enzyme conjugated anti-IgG polyclonal antibody is added and incubated at room temperature.

If toxin is present, a reactive antibody-enzyme complex is formed. After washing to remove unbound conjugate, substrate is added and incubated for 10 minutes at room temperature. Color develops in the presence of bound enzyme. Stop solution is added and the results are interpreted visually or spectrophotometrically.