Triapine ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
Synonyms: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
CAS Number:143621-35-6
:
Formula:C₇H₉N₅S
MDL Number:MFCD00924747
Molecular Weight:195.24 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable: