TileSTAT™ Interlocking ESD Rubber Anti-Fatigue Floor Mats, Transforming Technologies
Quickly snap together an ESD floor that can handle heavy traffic, and can be installed by anyone.
- 100% Nitrile rubber base compound specially formulated to drain static electricity
- Unique multi-nib support design offers exceptional fatigue relief and aeration
- Solid top design with an anti-slip surface texture
- Modular matting system allows for on-site custom configurations
TileSTAT™ ESD Anti-Fatigue Mats are a heavy-duty, durable safety mat constructed of conductive nitrile rubber for use in the harshest environments. TileSTAT™ mats feature a solid pebble embossed surface for improved traction. The foot interlocking mats fit together like a puzzle to create custom ESD workstations or complete room systems. Mats can be quickly dismantled to make moving and cleaning easy. Optional three inch wide beveled borders/side ramps can be added to the mats.
: Meets or exceeds requirements of ANSI ESD-S20.20 and ESD STM7.1